Product Details

SNP ID
rs4267608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:125961657 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTATAAATAAATAAGTACAATTT[A/G]CAAACCATTTGATTACATCCCTGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG1L PubMed Links

Gene Details

Gene
ALG1L
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015050.2 Intron NP_001015050.2
NM_001195223.1 Intron NP_001182152.1
XM_017005844.1 Intron XP_016861333.1
XM_017005845.1 Intron XP_016861334.1
XM_017005846.1 Intron XP_016861335.1
XM_017005847.1 Intron XP_016861336.1
Gene
ROPN1B
Gene Name
rhophilin associated tail protein 1B
There are no transcripts associated with this gene.

View Full Product Details