Product Details

SNP ID
rs12222448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46606983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGTACTTTTTAAAGATTATCA[C/G]GCTGGGCACGGTGGCTCACACCTGT
Phenotype
MIM: 615086
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HARBI1 PubMed Links

Gene Details

Gene
HARBI1
Gene Name
harbinger transposase derived 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173811.3 Intron NP_776172.1
XM_011520025.1 Intron XP_011518327.1
XM_011520026.2 Intron XP_011518328.1
XM_011520027.1 Intron XP_011518329.1
XM_011520028.2 Intron XP_011518330.1
XM_011520029.2 Intron XP_011518331.1
XM_017017602.1 Intron XP_016873091.1
XM_017017603.1 Intron XP_016873092.1
XM_017017604.1 Intron XP_016873093.1
XM_017017605.1 Intron XP_016873094.1
XM_017017606.1 Intron XP_016873095.1

View Full Product Details