Product Details

SNP ID
rs12991014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202206293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACTGAGTATTAAAAAGGTATTG[G/T]GGCCTTAATTAAATCCTTGATTCAG
Phenotype
MIM: 601912
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA2012 PubMed Links

Gene Details

Gene
KIAA2012
Gene Name
KIAA2012
There are no transcripts associated with this gene.

Gene
SUMO1
Gene Name
small ubiquitin-like modifier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005781.1 1339 UTR 3 NP_001005781.1
NM_001005782.1 1339 UTR 3 NP_001005782.1
NM_003352.4 1339 UTR 3 NP_003343.1

View Full Product Details