Product Details

SNP ID
hCV30823787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:151054904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGTGACCAGGACTAGAATCCCA[G/T]GTTTTTACTTCCCCATTGAGTATTT
Phenotype
MIM: 611275 MIM: 604684
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BNIPL PubMed Links

Gene Details

Gene
BNIPL
Gene Name
BCL2 interacting protein like
There are no transcripts associated with this gene.

Gene
C1orf56
Gene Name
chromosome 1 open reading frame 56
There are no transcripts associated with this gene.

Gene
CDC42SE1
Gene Name
CDC42 small effector 1
There are no transcripts associated with this gene.

Gene
MLLT11
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11
There are no transcripts associated with this gene.

View Full Product Details