Product Details

SNP ID
rs13315530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:193596628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCATTTCCATGAACTTTTAAGA[A/G]AACTCTTAAAGACATGTTTAATTCT
Phenotype
MIM: 605290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OPA1 PubMed Links

Gene Details

Gene
OPA1
Gene Name
OPA1, mitochondrial dynamin like GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015560.2 Intron NP_056375.2
NM_130831.2 Intron NP_570844.1
NM_130832.2 Intron NP_570845.1
NM_130833.2 Intron NP_570846.1
NM_130834.2 Intron NP_570847.2
NM_130835.2 Intron NP_570848.1
NM_130836.2 Intron NP_570849.2
NM_130837.2 Intron NP_570850.2
XM_017006500.1 Intron XP_016861989.1

View Full Product Details