Product Details

SNP ID
rs12525246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:12003743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTGTTTACTTCATTGTTAACAT[C/G]TTACATTACCAGGATACATTTATCA
Phenotype
MIM: 194540
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HIVEP1 PubMed Links

Gene Details

Gene
HIVEP1
Gene Name
human immunodeficiency virus type I enhancer binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002114.3 Intron NP_002105.3
XM_011514546.2 Intron XP_011512848.1
XM_011514547.2 Intron XP_011512849.1
XM_011514548.2 Intron XP_011512850.1
XM_011514549.2 Intron XP_011512851.1
XM_011514550.2 Intron XP_011512852.1
XM_011514551.2 Intron XP_011512853.1
XM_011514552.2 Intron XP_011512854.1
XM_011514553.1 Intron XP_011512855.1
XM_011514555.2 Intron XP_011512857.1
XM_017010800.1 Intron XP_016866289.1
XM_017010801.1 Intron XP_016866290.1
XM_017010802.1 Intron XP_016866291.1
XM_017010803.1 Intron XP_016866292.1
XM_017010804.1 Intron XP_016866293.1
Gene
LOC101928253
Gene Name
uncharacterized LOC101928253
There are no transcripts associated with this gene.

View Full Product Details