Product Details
- SNP ID
-
rs11950448
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:43360163 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGGCAATTACACTACCGGGTAAT[C/G]CTGCACAAATGAAAACTCAAATGTC
- Phenotype
-
MIM: 605240
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCL28
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77589834] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCL28
- Gene Name
- C-C motif chemokine ligand 28
View Full Product Details