Product Details

SNP ID
rs13241299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44104727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTGGCGTTGCTGGCCCTGTGCC[C/T]TGGAGGGCGCCCGCAGACGGTGCTG
Phenotype
MIM: 602981 MIM: 600815
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AEBP1 PubMed Links

Gene Details

Gene
AEBP1
Gene Name
AE binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129.4 367 Missense Mutation CCT,CTT P21L NP_001120.3
XM_011515162.1 367 Missense Mutation CCT,CTT P21L XP_011513464.1
Gene
MIR4649
Gene Name
microRNA 4649
There are no transcripts associated with this gene.

Gene
POLD2
Gene Name
DAN polymerase delta 2, accessory subunit
There are no transcripts associated with this gene.

View Full Product Details