Product Details

SNP ID
rs12090654
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:84645499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAACTCACTTTAAAATGGAAACA[C/T]GCAACAAAAAACAGATGCTGCTTTC
Phenotype
MIM: 608690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SSX2IP PubMed Links

Gene Details

Gene
SSX2IP
Gene Name
SSX family member 2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166293.1 4055 UTR 3 NP_001159765.1
NM_001166294.1 4055 UTR 3 NP_001159766.1
NM_001166295.1 4055 UTR 3 NP_001159767.1
NM_001166417.1 4055 UTR 3 NP_001159889.1
NM_014021.3 4055 UTR 3 NP_054740.3
XM_005270427.1 4055 UTR 3 XP_005270484.1
XM_005270428.1 4055 UTR 3 XP_005270485.1
XM_005270429.1 4055 UTR 3 XP_005270486.1
XM_006710331.1 4055 UTR 3 XP_006710394.1
XM_011540610.1 4055 UTR 3 XP_011538912.1
XM_011540612.2 4055 Intron XP_011538914.1
XM_017000234.1 4055 UTR 3 XP_016855723.1
XM_017000235.1 4055 UTR 3 XP_016855724.1
XM_017000236.1 4055 UTR 3 XP_016855725.1
XM_017000237.1 4055 UTR 3 XP_016855726.1

View Full Product Details