Product Details

SNP ID
rs7182613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66338269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCATCTAAAAAAACAAAACTG[A/G]AAACTACTAGAAAGAGGTCTAATAA
Phenotype
MIM: 614183 MIM: 610716
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIS3L PubMed Links

Gene Details

Gene
DIS3L
Gene Name
DIS3 like exosome 3'-5' exoribonuclease
There are no transcripts associated with this gene.

Gene
SCARNA14
Gene Name
small Cajal body-specific RNA 14
There are no transcripts associated with this gene.

Gene
TIPIN
Gene Name
TIMELESS interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289986.1 Intron NP_001276915.1
NM_017858.2 Intron NP_060328.2
XM_005254521.2 Intron XP_005254578.1
XM_005254522.4 Intron XP_005254579.1
XM_006720594.3 Intron XP_006720657.1
XM_011521743.1 Intron XP_011520045.1
XM_017022390.1 Intron XP_016877879.1
XM_017022391.1 Intron XP_016877880.1

View Full Product Details