Product Details

SNP ID
rs12433518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74713391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGAGGGTCTGGGTTATTTGAG[C/G]CAAGAAGGGAAGGCAATAGACAAGA
Phenotype
MIM: 615380
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AREL1 PubMed Links

Gene Details

Gene
AREL1
Gene Name
apoptosis resistant E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039479.1 Intron NP_001034568.1
XM_005268244.3 Intron XP_005268301.1
XM_006720344.2 Intron XP_006720407.1
XM_011537415.2 Intron XP_011535717.1
XM_011537416.2 Intron XP_011535718.1
XM_011537417.2 Intron XP_011535719.1
Gene
FCF1
Gene Name
FCF1 rRNA-processing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318508.1 Intron NP_001305437.1
NM_015962.4 Intron NP_057046.1
XM_005267730.3 Intron XP_005267787.1
XM_011536815.2 Intron XP_011535117.1
XM_011536816.2 Intron XP_011535118.1

View Full Product Details