Product Details

SNP ID
rs12191154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111663112 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTTTTGCTTCCCTTTTTCATTGT[A/G]TCACCTTCTGCCAGCTCCCCGTCAA
Phenotype
MIM: 137025
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FYN PubMed Links

Gene Details

Gene
FYN
Gene Name
FYN proto-oncogene, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002037.5 Intron NP_002028.1
NM_153047.3 Intron NP_694592.1
NM_153048.3 Intron NP_694593.1
XM_005266890.3 Intron XP_005266947.1
XM_005266892.3 Intron XP_005266949.1
XM_017010650.1 Intron XP_016866139.1
XM_017010651.1 Intron XP_016866140.1
XM_017010652.1 Intron XP_016866141.1
XM_017010653.1 Intron XP_016866142.1
XM_017010654.1 Intron XP_016866143.1
XM_017010655.1 Intron XP_016866144.1

View Full Product Details