Product Details
- SNP ID
-
rs12812693
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:92739438 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGGGAGTATTGCAGGGGGTGGTA[A/C]CAATAAATGTTCACCGCAGAGAACG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PLEKHG7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116636805] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PLEKHG7
- Gene Name
- pleckstrin homology and RhoGEF domain containing G7
View Full Product Details