Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001297741.1 | Intron | NP_001284670.1 | |||
NM_001297742.1 | Intron | NP_001284671.1 | |||
NM_032320.6 | Intron | NP_115696.2 | |||
XM_017018405.1 | Intron | XP_016873894.1 | |||
XM_017018406.1 | Intron | XP_016873895.1 | |||
XM_017018407.1 | Intron | XP_016873896.1 | |||
XM_017018408.1 | Intron | XP_016873897.1 | |||
XM_017018409.1 | Intron | XP_016873898.1 | |||
XM_017018410.1 | Intron | XP_016873899.1 | |||
XM_017018411.1 | Intron | XP_016873900.1 |