Product Details

SNP ID
rs4910657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4645071 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTAAGAGAGGCTTTTTCTGACC[C/G]CTGTCTCCAGTATGAGTGTCCTGCA
Phenotype
MIM: 611267
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR51D1 PubMed Links

Gene Details

Gene
OR51D1
Gene Name
olfactory receptor family 51 subfamily D member 1
There are no transcripts associated with this gene.

Gene
OR51E1
Gene Name
olfactory receptor family 51 subfamily E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152430.3 Intron NP_689643.2

View Full Product Details