Product Details

SNP ID
rs10845175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10591119 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTGAGGTCTTAAATTCACCACCC[A/G]TTGGTGATACCCATTCTGGCCTCTG
Phenotype
MIM: 604274
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLRA1P PubMed Links

Gene Details

Gene
KLRA1P
Gene Name
killer cell lectin like receptor A1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details