Product Details

SNP ID
rs11795808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73945448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAAATTTTGGTACATAAATATTA[A/T]GAAATATTTTGACTTTTGACCTAGA
Phenotype
MIM: 300832
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
JPX PubMed Links

Gene Details

Gene
JPX
Gene Name
JPX transcript, XIST activator (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details