Product Details

SNP ID
rs11795610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:75381460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTCCTCTCAATTTATAGTGTTA[C/G]TACCACTCATGAATCCATTGCAACA
Phenotype
MIM: 300576
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZDHHC15 PubMed Links

Gene Details

Gene
ZDHHC15
Gene Name
zinc finger DHHC-type containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146256.1 Intron NP_001139728.1
NM_001146257.1 Intron NP_001139729.1
NM_144969.2 Intron NP_659406.1
XM_006724624.3 Intron XP_006724687.1
XM_017029296.1 Intron XP_016884785.1
XM_017029297.1 Intron XP_016884786.1
XM_017029298.1 Intron XP_016884787.1

View Full Product Details