Product Details

SNP ID
rs12442178
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:39599198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTATTTTTAATAACACTTTCTAT[A/C]TAAATCAGGGTGACTTTTTAAAAAA
Phenotype
MIM: 615795 MIM: 188060
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
FSIP1 PubMed Links

Gene Details

Gene
FSIP1
Gene Name
fibrous sheath interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324338.1 2947 Intron NP_001311267.1
NM_152597.4 2947 Intron NP_689810.3
XM_011521305.2 2947 UTR 3 XP_011519607.1
XM_011521306.1 2947 Intron XP_011519608.1
XM_011521307.2 2947 Intron XP_011519609.1
XM_011521309.2 2947 Intron XP_011519611.1
XM_011521311.2 2947 Intron XP_011519613.1
XM_017021972.1 2947 Intron XP_016877461.1
Gene
THBS1
Gene Name
thrombospondin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003246.3 2947 Intron NP_003237.2
XM_011521971.1 2947 Intron XP_011520273.1

View Full Product Details