Product Details

SNP ID
rs12906047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42069803 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTGGATGGGCCTAGCCTGAAGCC[C/T]TCTCCTGGCAGACTCATTTCCCTTC
Phenotype
MIM: 612864
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G4D PubMed Links

Gene Details

Gene
PLA2G4D
Gene Name
phospholipase A2 group IVD
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178034.3 Intron NP_828848.3
XM_011521467.2 Intron XP_011519769.1

View Full Product Details