Product Details

SNP ID
rs6505173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30319039 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAACCTTCTCAACAATCTCAAAA[A/G]GTAGGTATGATTGTCCTCATTTCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMIGD1 PubMed Links

Gene Details

Gene
TMIGD1
Gene Name
transmembrane and immunoglobulin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319942.1 Intron NP_001306871.1
NM_206832.2 Intron NP_996663.1
XM_011524787.1 Intron XP_011523089.1

View Full Product Details