Product Details

SNP ID
rs13174941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:661381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATAGAACCTGTCCCTGTGTCCT[C/G]GTGTCACCCATGACAGAACCTGTCC
Phenotype
MIM: 616475 MIM: 608773
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEP72 PubMed Links

Gene Details

Gene
CEP72
Gene Name
centrosomal protein 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018140.3 6699 Intron NP_060610.2
XM_005248322.3 6699 Intron XP_005248379.1
XM_011514063.1 6699 Intron XP_011512365.1
XM_011514064.2 6699 Intron XP_011512366.1
XM_017009626.1 6699 Intron XP_016865115.1
XM_017009627.1 6699 Intron XP_016865116.1
Gene
LOC101929895
Gene Name
uncharacterized LOC101929895
There are no transcripts associated with this gene.

Gene
TPPP
Gene Name
tubulin polymerization promoting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007030.2 6699 UTR 3 NP_008961.1
XM_005248237.3 6699 UTR 3 XP_005248294.2
XM_017008993.1 6699 UTR 3 XP_016864482.1
XM_017008994.1 6699 UTR 3 XP_016864483.1

View Full Product Details