Product Details
- SNP ID
-
rs11882699
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39708134 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTCAGAGGAAATCTTTTCCTATG[C/G]AAGCATGTAGATATGTCTATGATTG
- Phenotype
-
MIM: 607260
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LGALS14
PubMed Links
Gene Details
- Gene
- LGALS14
- Gene Name
- galectin 14
View Full Product Details