Product Details

SNP ID
rs12985291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56088471 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCTCGCCATCGCCCACGGGGATG[G/T]CGATCTCGCCGTCCGCCGCCACCGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928886 PubMed Links

Gene Details

Gene
LOC101928886
Gene Name
uncharacterized LOC101928886
There are no transcripts associated with this gene.

Gene
ZNF787
Gene Name
zinc finger protein 787
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002836.3 913 Missense Mutation GAC,GCC D234A NP_001002836.2
XM_011526445.2 913 Missense Mutation GAC,GCC D234A XP_011524747.1

View Full Product Details