Product Details
- SNP ID
-
rs13438302
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:78019354 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGAGGCGCCGGGTTTGAGGACGCT[C/G]GGCAGCTTGTCAGAACCCGGCACCT
- Phenotype
-
MIM: 606382
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MAGI2
PubMed Links
Gene Details
- Gene
- MAGI2
- Gene Name
- membrane associated guanylate kinase, WW and PDZ domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301128.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1429P |
NP_001288057.1 |
NM_012301.3 |
5069 |
Silent Mutation |
CCC,CCG |
P1443P |
NP_036433.2 |
XM_011516718.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1485P |
XP_011515020.1 |
XM_011516719.2 |
5069 |
Silent Mutation |
CCC,CCG |
P1365P |
XP_011515021.1 |
XM_011516720.2 |
5069 |
Silent Mutation |
CCC,CCG |
P1365P |
XP_011515022.1 |
XM_011516726.2 |
5069 |
Silent Mutation |
CCC,CCG |
P1137P |
XP_011515028.1 |
XM_011516728.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1094P |
XP_011515030.1 |
XM_017012840.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1528P |
XP_016868329.1 |
XM_017012841.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1527P |
XP_016868330.1 |
XM_017012842.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1526P |
XP_016868331.1 |
XM_017012843.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1514P |
XP_016868332.1 |
XM_017012844.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1486P |
XP_016868333.1 |
XM_017012845.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1483P |
XP_016868334.1 |
XM_017012846.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1471P |
XP_016868335.1 |
XM_017012847.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1365P |
XP_016868336.1 |
XM_017012848.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1322P |
XP_016868337.1 |
XM_017012849.1 |
5069 |
Silent Mutation |
CCC,CCG |
P1320P |
XP_016868338.1 |
XM_017012850.1 |
5069 |
Intron |
|
|
XP_016868339.1 |
XM_017012851.1 |
5069 |
Intron |
|
|
XP_016868340.1 |
XM_017012852.1 |
5069 |
Intron |
|
|
XP_016868341.1 |
View Full Product Details