Product Details

SNP ID
rs13032333
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9480572 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTTCAGTGCAAACTCATTTTTT[A/T]AAAAGTGTATGTATCTTCATGAAAA
Phenotype
MIM: 603639 MIM: 606029
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ADAM17 PubMed Links
Additional Information
For this assay, SNP(s) [rs79638552] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ADAM17
Gene Name
ADAM metallopeptidase domain 17
There are no transcripts associated with this gene.

Gene
CPSF3
Gene Name
cleavage and polyadenylation specific factor 3
There are no transcripts associated with this gene.

Gene
IAH1
Gene Name
isoamyl acetate-hydrolyzing esterase 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039613.2 Intron NP_001034702.1
NM_001320858.1 Intron NP_001307787.1
NM_001320859.1 Intron NP_001307788.1
NM_001320860.1 Intron NP_001307789.1
NM_001320863.1 Intron NP_001307792.1
XM_017003878.1 Intron XP_016859367.1

View Full Product Details