Product Details

SNP ID
rs6480448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70258487 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCCTAGGTTGTGATTAGATAAAT[G/T]CATGAATGACCAGAGGGGAAGTTAC
Phenotype
MIM: 607448
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NPFFR1 PubMed Links

Gene Details

Gene
NPFFR1
Gene Name
neuropeptide FF receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022146.4 Intron NP_071429.1

View Full Product Details