Product Details

SNP ID
rs10999769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71320878 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGATGCCTGTACTTGGAACCAG[C/T]AGACACAGATTCCCATTCCCAGCCA
Phenotype
MIM: 612373
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC29A3 PubMed Links

Gene Details

Gene
SLC29A3
Gene Name
solute carrier family 29 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174098.1 Intron NP_001167569.1
NM_018344.5 Intron NP_060814.4
XM_006717910.3 Intron XP_006717973.1
XM_017016377.1 Intron XP_016871866.1
XM_017016378.1 Intron XP_016871867.1

View Full Product Details