Product Details

SNP ID
rs10898973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74264988 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAAGCACAAGATTTGGGGCCAG[G/T]CTGTGGCACTCTCAACAAGTGACTT
Phenotype
MIM: 608987
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
P4HA3 PubMed Links

Gene Details

Gene
P4HA3
Gene Name
prolyl 4-hydroxylase subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288748.1 Intron NP_001275677.1
NM_182904.4 Intron NP_878907.1

View Full Product Details