Product Details

SNP ID
rs11257347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:11827298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCGTTGCTGGACATCTCTTCA[A/G]ATGCTTGTTGGCCACATCTTCTTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PROSER2 PubMed Links

Gene Details

Gene
PROSER2
Gene Name
proline and serine rich 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153256.3 Intron NP_694988.3
XM_011519438.2 Intron XP_011517740.1
XM_017016028.1 Intron XP_016871517.1

View Full Product Details