Product Details
- SNP ID
-
rs10906025
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:11827247 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAGTGGGTATGAAGTAGTATCTCA[C/T]GGTGGTTTTGGTGCATTTCTCGAAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PROSER2
PubMed Links
Gene Details
- Gene
- PROSER2
- Gene Name
- proline and serine rich 2
View Full Product Details