Product Details
- SNP ID
-
rs11101291
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:48317552 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTCTTTTAAGATTAGCTCTGAAGT[A/G]AACAAAGAAATGGGTTGGTGGATAA
- Phenotype
-
MIM: 601158
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MAPK8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142886685] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAPK8
- Gene Name
- mitogen-activated protein kinase 8
View Full Product Details