Product Details

SNP ID
rs10877854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:62472178 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGAAAGCCAATAGTTTGGATTGA[C/T]TGGATGATGATGGGAAGGTTAAAGG
Phenotype
MIM: 616822
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MON2 PubMed Links

Gene Details

Gene
MON2
Gene Name
MON2 homolog, regulator of endosome-to-Golgi trafficking
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278469.1 Intron NP_001265398.1
NM_001278470.1 Intron NP_001265399.1
NM_001278471.1 Intron NP_001265400.1
NM_001278472.1 Intron NP_001265401.1
NM_015026.2 Intron NP_055841.2
XM_017019041.1 Intron XP_016874530.1
XM_017019042.1 Intron XP_016874531.1
XM_017019043.1 Intron XP_016874532.1
XM_017019044.1 Intron XP_016874533.1
XM_017019045.1 Intron XP_016874534.1
XM_017019046.1 Intron XP_016874535.1

View Full Product Details