Product Details

SNP ID
hCV31979918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102136409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGCAGAAGAGGAAAGGGAAGGT[A/C]TGGCCTGTCTGTATGTGATCCTGTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPRC1 PubMed Links

Gene Details

Gene
PPRC1
Gene Name
peroxisome proliferator-activated receptor gamma, coactivator-related 1
There are no transcripts associated with this gene.

View Full Product Details