Product Details

SNP ID
rs12293021
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35139292 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCTTTCGCCCGCGCCCTCCGTTC[A/G]CTCCGGACACCATGGACAAGTTTTG
Phenotype
MIM: 107269
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CD44 PubMed Links

Gene Details

Gene
CD44
Gene Name
CD44 molecule (Indian blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000610.3 150 UTR 5 NP_000601.3
NM_001001389.1 150 UTR 5 NP_001001389.1
NM_001001390.1 150 UTR 5 NP_001001390.1
NM_001001391.1 150 UTR 5 NP_001001391.1
NM_001001392.1 150 UTR 5 NP_001001392.1
NM_001202555.1 150 UTR 5 NP_001189484.1
NM_001202556.1 150 UTR 5 NP_001189485.1
NM_001202557.1 150 UTR 5 NP_001189486.1
XM_005253231.2 150 UTR 5 XP_005253288.1
XM_005253232.2 150 UTR 5 XP_005253289.1
XM_005253233.2 150 UTR 5 XP_005253290.1
XM_005253234.2 150 UTR 5 XP_005253291.1
XM_005253235.2 150 UTR 5 XP_005253292.1
XM_005253238.2 150 UTR 5 XP_005253295.1
XM_005253239.2 150 UTR 5 XP_005253296.1
XM_005253240.2 150 UTR 5 XP_005253297.1
XM_006718388.1 150 UTR 5 XP_006718451.1
XM_006718390.3 150 UTR 5 XP_006718453.1
XM_011520482.1 150 UTR 5 XP_011518784.1
XM_011520483.1 150 UTR 5 XP_011518785.1
XM_011520484.1 150 UTR 5 XP_011518786.1
XM_011520485.1 150 UTR 5 XP_011518787.1
XM_011520486.1 150 UTR 5 XP_011518788.1
XM_011520487.2 150 UTR 5 XP_011518789.1
XM_011520488.1 150 UTR 5 XP_011518790.1
XM_011520489.2 150 UTR 5 XP_011518791.1
XM_017018583.1 150 UTR 5 XP_016874072.1
XM_017018584.1 150 UTR 5 XP_016874073.1
XM_017018585.1 150 UTR 5 XP_016874074.1
Gene
LOC100507144
Gene Name
uncharacterized LOC100507144
There are no transcripts associated with this gene.

View Full Product Details