Product Details

SNP ID
rs13376803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43385907 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTCATCATAAGAATATAAGGGAG[A/G]TCATAGATTTGATGTATGAAATTTT
Phenotype
MIM: 616926 MIM: 601037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FXYD4 PubMed Links

Gene Details

Gene
FXYD4
Gene Name
FXYD domain containing ion transport regulator 4
There are no transcripts associated with this gene.

Gene
HNRNPF
Gene Name
heterogeneous nuclear ribonucleoprotein F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098204.1 2386 UTR 3 NP_001091674.1
NM_001098205.1 2386 UTR 3 NP_001091675.1
NM_001098206.1 2386 UTR 3 NP_001091676.1
NM_001098207.1 2386 UTR 3 NP_001091677.1
NM_001098208.1 2386 UTR 3 NP_001091678.1
NM_004966.3 2386 UTR 3 NP_004957.1

View Full Product Details