Product Details

SNP ID
rs9557425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:87672205 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAGTTGCCCCCTATGCAGATGG[C/G]AACTGTGAGTACCCCTGTGGGGGGG
Phenotype
MIM: 609680
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR4500HG PubMed Links

Gene Details

Gene
MIR4500HG
Gene Name
MIR4500 host gene
There are no transcripts associated with this gene.

Gene
SLITRK5
Gene Name
SLIT and NTRK like family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015567.1 878 Intron NP_056382.1
XM_005254038.4 878 UTR 5 XP_005254095.1
XM_005254039.2 878 Intron XP_005254096.1

View Full Product Details