Product Details

SNP ID
rs4242878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8173064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGACTTAGGATTGTGATGAATGCA[A/G]GCGATAGAGATGAGCGAAGAGCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM66C PubMed Links

Gene Details

Gene
FAM66C
Gene Name
family with sequence similarity 66 member C
There are no transcripts associated with this gene.

Gene
ZNF705A
Gene Name
zinc finger protein 705A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004328.2 Intron NP_001004328.1

View Full Product Details