Product Details
- SNP ID
-
rs4266605
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:81443917 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAACATTCGGAAGAGTACATGAAT[C/T]TATTTACATTCAATGTATGTGTTAA
- Phenotype
-
MIM: 170715
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PMP2
PubMed Links
Gene Details
- Gene
- PMP2
- Gene Name
- peripheral myelin protein 2
There are no transcripts associated with this gene.
View Full Product Details