Product Details

SNP ID
rs3213559
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100826600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAGTATATTGAGTATCCACAGAG[A/G]CATATACATACTCTGTTTTTTTTGC
Phenotype
MIM: 300907
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CSTF2 PubMed Links

Gene Details

Gene
CSTF2
Gene Name
cleavage stimulation factor subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306206.1 Intron NP_001293135.1
NM_001306209.1 Intron NP_001293138.1
NM_001325.2 Intron NP_001316.1

View Full Product Details