Product Details

SNP ID
rs34140799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7380749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGGGCGCTTATTCATGTAAGAGG[T/G]AGGGTCCTGGCTGAGAGGTGAAGGA
Phenotype
MIM: 607611 MIM: 608076
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
PLSCR3 PubMed Links

Gene Details

Gene
PLSCR3
Gene Name
phospholipid scramblase 3
There are no transcripts associated with this gene.

Gene
TMEM256-PLSCR3
Gene Name
TMEM256-PLSCR3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TNK1
Gene Name
tyrosine kinase non receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251902.1 Intron NP_001238831.1
NM_003985.4 Intron NP_003976.2
XM_011524045.2 Intron XP_011522347.1
XM_011524046.2 Intron XP_011522348.1
XM_011524047.2 Intron XP_011522349.1
XM_011524048.1 Intron XP_011522350.1

View Full Product Details