Product Details

SNP ID
rs7893855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72222970 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGATTAAACACTCACAGGTAGGT[G/T]CTCTGTTCATAATAGGCCATGGTTT
Phenotype
MIM: 613427 MIM: 614215
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANAPC16 PubMed Links

Gene Details

Gene
ANAPC16
Gene Name
anaphase promoting complex subunit 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242546.1 Intron NP_001229475.1
NM_001242547.1 Intron NP_001229476.1
NM_001242548.1 Intron NP_001229477.1
NM_173473.3 Intron NP_775744.1
Gene
ASCC1
Gene Name
activating signal cointegrator 1 complex subunit 1
There are no transcripts associated with this gene.

View Full Product Details