Product Details

SNP ID
rs16853981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:42111873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTTTGCACTAGGAACCTATGG[A/G]ACAAATGTGAACAAACTCGTGATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BEND4 PubMed Links

Gene Details

Gene
BEND4
Gene Name
BEN domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159547.1 7830 UTR 3 NP_001153019.1
NM_207406.3 7830 UTR 3 NP_997289.2
XM_017008185.1 7830 UTR 3 XP_016863674.1

View Full Product Details