Product Details

SNP ID
rs17199923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6688522 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTGAGAAACCTCCATAGGTATCA[A/T]TTCCACAAATATGCTATGAATATAG
Phenotype
MIM: 194536
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF12 PubMed Links

Gene Details

Gene
ZNF12
Gene Name
zinc finger protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006956.2 4860 UTR 3 NP_008887.2
NM_016265.3 4860 UTR 3 NP_057349.2

View Full Product Details