Product Details

SNP ID
rs17089375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74540755 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGGATTTTCAGGGTGATTGGCGA[A/C]GTTGTGGCCATGAGTGCTCGGCTCA
Phenotype
MIM: 609064
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNDP1 PubMed Links

Gene Details

Gene
CNDP1
Gene Name
carnosine dipeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032649.5 Intron NP_116038.4

View Full Product Details