Product Details

SNP ID
rs16954159
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4734254 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAAATACACATAGAATTTGTCCA[A/G]GTTATTATCACCCAGTGTGAAAAGC
Phenotype
MIM: 605398 MIM: 612383 MIM: 614312
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CXCL16 PubMed Links
Additional Information
For this assay, SNP(s) [rs11289101] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CXCL16
Gene Name
C-X-C motif chemokine ligand 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100812.1 1603 Intron NP_001094282.1
NM_022059.3 1603 UTR 3 NP_071342.2
Gene
MED11
Gene Name
mediator complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001683.3 1603 Intron NP_001001683.1
NM_001305000.1 1603 Intron NP_001291929.1
Gene
ZMYND15
Gene Name
zinc finger MYND-type containing 15
There are no transcripts associated with this gene.

View Full Product Details