Product Details

SNP ID
rs17700166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:38724443 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGAAATGCTGAGAAGGCTGAGG[A/G]GTACATTCAATAATAACAAAGAGCC
Phenotype
MIM: 605485
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS41 PubMed Links

Gene Details

Gene
VPS41
Gene Name
VPS41, HOPS complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014396.3 4319 UTR 3 NP_055211.2
NM_080631.3 4319 UTR 3 NP_542198.2
XM_017011988.1 4319 Intron XP_016867477.1

View Full Product Details