Product Details

SNP ID
rs17712249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:47215718 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCCCTAGCTGCGTGTGACTGCAG[A/C]ATTCTGGAAAAACAGCAGAGGGCAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SKOR2 PubMed Links

Gene Details

Gene
SKOR2
Gene Name
SKI family transcriptional corepressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037802.1 Intron NP_001032891.1
NM_001278063.1 Intron NP_001264992.1
XM_006722525.3 Intron XP_006722588.1

View Full Product Details