Product Details
- SNP ID
-
rs17675862
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
31
- Location
-
Chr.1:157678489 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTGCCTTGCCACGTGTCTCCACT[A/G]TGAAAATAACACAGTGCTTGCTCAG
- Phenotype
-
MIM: 606510
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FCRL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16838952] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FCRL3
- Gene Name
- Fc receptor like 3
View Full Product Details