Product Details

SNP ID
rs17078556
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:7038624 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGGATGAAGCAGGTCCCACCAA[C/G]ATGTTCTCCAAAAATGCAGTATAGT
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
DEFA11P PubMed Links
Additional Information
For this assay, SNP(s) [rs115835187,rs192288533] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DEFA11P
Gene Name
defensin alpha 11, pseudogene
There are no transcripts associated with this gene.

Gene
DEFA7P
Gene Name
defensin alpha 7, pseudogene
There are no transcripts associated with this gene.

View Full Product Details